Please login to your online account to display your discounted pricing

(-)-Cytisine, Tocris

Potent, selective neuronal nicotinic agonist

$113.57 - $449.15

Chemical Identifiers

Chemical Name or Material (-)-Cytisine
Assay Percent Range >99%
Molecular Formula C11H14N2O
Formula Weight 190.24
Synonym (1R,5S)-1,2,3,4,5,6-hexahydro-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

139010

 
r & d systems
RDS139010
10mg Each for $113.57

139050

 
r & d systems
RDS139050
50mg Each for $449.15
Description & Specifications

Specifications

Formula Weight 190.24
Quantity 10mg
Molecular Formula C11H14N2O
Assay Percent Range >99%
Chemical Name or Material (-)-Cytisine
Synonym (1R,5S)-1,2,3,4,5,6-hexahydro-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one
CAS 485-35-8
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature