missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytidine, 99+%, ACROS Organics™

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.219
MDL Number MFCD00006545
InChI Key UHDGCWIWMRVCDJ-XVFCMESISA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC111810100 SDS View Documents Acros Organics
111810100
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC111810500 SDS View Documents Acros Organics
111810500
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC111812500 SDS View Documents Acros Organics
AC111812500
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

65-46-3
243.219
UHDGCWIWMRVCDJ-XVFCMESISA-N
6175
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
C9H13N3O5
MFCD00006545
cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
CHEBI:17562
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O

Specifications

0.43 to 0.47 (A250/A260), 2.05 to 2.15 (A280/A260)
0.5% max. (105°C, 5 hrs)
+ 31.50
0.1% max. (800°C)
White
10g
99+%
MFCD00006545
UHDGCWIWMRVCDJ-XVFCMESISA-N
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
6175
243.22
≥99%
Authentic
Glass bottle
98 to 102% (lambda max. 280nm, pH 1) molar absorptivity 13000 +/- 2%, on dry basis
98% min. (c=1, H2O, 430nm) 1cm cell
210.0°C to 217.0°C
65-46-3
C9H13N3O5
cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O
243.219
CHEBI:17562
Crystalline Powder
Cytidine, 99.5%
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 200-610-9

RTECSNumber : UW7370000

TSCA : TSCA