missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytidine 5'-triphosphate, disodium salt dihydrate, 95%, ACROS Organics™

$466.92 - $466.92

Chemical Identifiers

CAS 81012-87-5
Molecular Formula C9H16N3Na4O14P3
Molecular Weight (g/mol) 575.114
MDL Number MFCD00150807
InChI Key LDTPLTXNLOEHLN-ODQFIEKDSA-N
Synonym cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
PubChem CID 129893565
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC226225000 SDS View Documents Acros Organics
226225000
500mg Each for $466.92
Specifications

Chemical Identifiers

81012-87-5
575.114
LDTPLTXNLOEHLN-ODQFIEKDSA-N
129893565
C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
C9H16N3Na4O14P3
MFCD00150807
cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium

Specifications

Authentic
8.5% max. (K.F.)
215.0°C to 218.0°C
81012-87-5
C9H16N3Na4O14P3
cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
575.114
563.15
95%
Glass bottle
White
500mg
95%
MFCD00150807
LDTPLTXNLOEHLN-ODQFIEKDSA-N
[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
129893565
Crystalline Powder
Cytidine 5'-triphosphate, disodium salt dihydrate, 95%