missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cytidine-5'-diphosphate trisodium salt

$49.20 - $331.55

Chemical Identifiers

CAS 34393-59-4
Molecular Formula C9H21N3Na3O11P2
Molecular Weight (g/mol) 478.194
MDL Number MFCD00065201
InChI Key ZZTAOPCXVZSTKK-AVTKVCAJSA-N
Synonym cytidine 5'-diphosphate trisodium salt
PubChem CID 131674171
IUPAC Name [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium
SMILES [HH].[HH].[HH].C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64350MC View Documents Alfa Aesar™
J64350MC
100mg Each for $49.20
 
AAJ64350ME View Documents Alfa Aesar™
J64350ME
500mg Each for $197.39
 
AAJ6435003 View Documents Alfa Aesar™
J6435003
1g Each for $331.55
 
Description

Description

Applications
Cytidine-5′-diphosphate trisodium salt has been used to produce a vaccinia virus that induces ribonucleotide reductase in primate cells.

Solubility
Freely soluble in water, insoluble in ethanol, chloroform and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

34393-59-4
478.194
ZZTAOPCXVZSTKK-AVTKVCAJSA-N
131674171
[HH].[HH].[HH].C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na]
C9H21N3Na3O11P2
MFCD00065201
cytidine 5'-diphosphate trisodium salt
[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium

Specifications

White
34393-59-4
MFCD00065201
ZZTAOPCXVZSTKK-AVTKVCAJSA-N
[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium
131674171
Crystalline Powder
100mg
C9H21N3Na3O11P2
cytidine 5'-diphosphate trisodium salt
[HH].[HH].[HH].C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na]
478.194
469.12
Cytidine-5'-diphosphate trisodium salt
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C