Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS1249PABX

 View more versions of this product

Catalog No. MS1249PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Isotype IgG1
Regulatory Status RUO
Format Concentrated
Quantity 200µL
Immunogen Recombinant protein encoding NBF2 domain of human CFTR
Clone M3A7
Antigen Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4
Host Species Mouse
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4, Mouse Monoclonal Antibody.

CFTR (cystic fibrosis transmembrane conductance regulator) is composed of two membrane-spanning domains (MSD), two nucleotide-binding domains (NBD), and an R domain. It is structurally similar to multidrug resistance (Mdr1) protein and both are members of the superfamily of ATP-binding cassette (ABC) transporters, also known as traffic ATPases, which are implicated in the movement of various substrates. The CFTR protein is a small conductance adenosine 3',5'-cyclic monophosphate (cAMP)-activated chloride ion channel found in the apical membranes of epithelia within the pancreas, airway, intestine, bile duct, sweat gland, and male genital ducts. CFTR is a valuable marker of human pancreatic duct cell development and differentiation.

Host Species: Mouse

Clone: M3A7

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1370-1380

Immunogen: Recombinant protein encoding NBF2 domain of human CFTR

Molecular Weight: 165-170 kDa

Positive Control: Pancreas

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO