Learn More
Cystamine Dihydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 111770250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cystamine dihydrochloride | |
220°C | |
Authentic | |
Plastic bottle | |
S2(CH2CH2NH2)2·2HCl | |
25 g | |
15, 2773 | |
Solubility in water: soluble. Other solubilities: slightly soluble in acetone,ethanol | |
C(CSSCCN)N.Cl.Cl | |
225.2 | |
225.2 | |
Crystalline Powder, Crystals or Chunks |
56-17-7 | |
White to Yellow | |
96% min. (Argentometry) | |
C4H12N2S2·2HCl | |
MFCD00012905 | |
04, 287 | |
cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride | |
YUFRRMZSSPQMOS-UHFFFAOYSA-N | |
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride | |
5941 | |
97% |
Chemical Identifiers
56-17-7 | |
225.2 | |
YUFRRMZSSPQMOS-UHFFFAOYSA-N | |
5941 | |
C(CSSCCN)N.Cl.Cl |
C4H12N2S2·2HCl | |
MFCD00012905 | |
cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride | |
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-260-7
RUO – Research Use Only