Please login to your online account to display your discounted pricing

Alfa Aesar™ [Cys&superscript_3;,&superscript_6;, Tyr&superscript_8;, Pro&superscript_1;&superscript_0;] Substance P

For cellular and molecular biology applications

Specifications

Product Type [Cys3,6, Tyr8, Pro10] Substance P
Molecular Weight 1295.56
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Cys-Pro-Gln-Cys-Phe-Tyr-Gly-Pro-Met-NH2, (Disulfide bridge: Cys3-Cys6)
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66549MA

 
alfa aesar™
J66549MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66549LB0

 
alfa aesar™
J66549LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Cys3,6, Tyr8, Pro10] Substance P
Molecular Weight 1295.56
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Cys-Pro-Gln-Cys-Phe-Tyr-Gly-Pro-Met-NH2, (Disulfide bridge: Cys3-Cys6)
Format Solid
Regulatory Status RUO
Purification Method HPLC