Please login to your online account to display your discounted pricing

Alfa Aesar™ Cys-Kemptide

For cellular and molecular biology applications

Specifications

Product Type Cys-Kemptide
Molecular Weight 875.06
Storage Requirements -30° to -10°C
Sequence Cys-Leu-Arg-Arg-Ala-Ser-Leu-Gly
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66384LB0

 
alfa aesar™
J66384LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66384MCR

 
alfa aesar™
J66384MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Cys-Kemptide
Molecular Weight 875.06
Storage Requirements -30° to -10°C
Sequence Cys-Leu-Arg-Arg-Ala-Ser-Leu-Gly
Format Solid
Regulatory Status RUO
Purification Method HPLC