missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyproheptadine Hydrochloride Sesquihydrate, TCI America™

Manufacturer:  TCI America C321825G

 View more versions of this product

Catalog No. C321825G



Specifications

Specifications

Cyproheptadine Hydrochloride Sesquihydrate
25g
C42H50Cl2N2O3
2811
ZEAUHIZSRUAMQG-UHFFFAOYSA-N
4-(dibenzo[1,2-a
74764338
323.86
≥98.0% (HPLC,T)
White-Yellow
41354-29-4
MFCD00012538
4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methylpiperidine Hydrochloride
[H+].[H+].CN1CCC(=C2C3=CC=CC=C3C=CC4=CC=CC=C42)CC1.CN1CCC(=C2C3=CC=CC=C3C=CC4=CC=CC=C42)CC1.O.O.O.[Cl-].[Cl-]
701.773
CHEBI:650917
Crystal-Powder at 20°C

Chemical Identifiers

41354-29-4
701.773
ZEAUHIZSRUAMQG-UHFFFAOYSA-N
74764338
4-(dibenzo[1,2-a
C42H50Cl2N2O3
MFCD00012538
4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methylpiperidine Hydrochloride
CHEBI:650917
[H+].[H+].CN1CCC(=C2C3=CC=CC=C3C=CC4=CC=CC=C42)CC1.CN1CCC(=C2C3=CC=CC=C3C=CC4=CC=CC=C42)CC1.O.O.O.[Cl-].[Cl-]
Safety and Handling

Safety and Handling

RTECSNumber : TM7050000

TSCA : No

Documents