Learn More
Cyproconazole, Thermo Scientific Chemicals
Triazole fungicide which causes hepatocellular adenomas and carcinomas
Supplier: Thermo Scientific Chemicals J6314209
Description
Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.
Solubility
Soluble in acetone, ethanol, xylene, and dimethyl sulfoxide.
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications
Cyproconazole | |
Crystalline | |
108°C | |
MFCD01678672 | |
8396421 | |
Soluble in acetone,ethanol,xylene,and dimethyl sulfoxide. | |
CC(C1CC1)C(CN2C=NC=N2)(C3=CC=C(C=C3)Cl)O | |
291.779 | |
CHEBI:83748 | |
Brown | |
>100°C (212°F) |
Cyproconazole | |
94361-06-5 | |
C15H18ClN3O | |
UN3077 | |
14,2771 | |
UFNOUKDBUJZYDE-UHFFFAOYSA-N | |
2-(4-chlorophenyl)-3-cyclopropyl-1-(1,2,4-triazol-1-yl)butan-2-ol | |
86132 | |
291.78 | |
10 g | |
>250°C |
Chemical Identifiers
94361-06-5 | |
291.779 | |
UFNOUKDBUJZYDE-UHFFFAOYSA-N | |
CHEBI:83748 | |
CC(C1CC1)C(CN2C=NC=N2)(C3=CC=C(C=C3)Cl)O |
C15H18ClN3O | |
MFCD01678672 | |
86132 | |
2-(4-chlorophenyl)-3-cyclopropyl-1-(1,2,4-triazol-1-yl)butan-2-ol |
Safety and Handling
GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301-H360D-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECSNumber : XZ4803250
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only