Please login to your online account to display your discounted pricing

CyPPA, Tocris

Activator of KCa2.2 and KCa2.3 channels

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material CyPPA
Assay Percent Range >99%
Molecular Formula C16H23N5
Formula Weight 285.39
Synonym N-Cyclohexyl-N-[2-(3, 5-dimethyl-pyrazol-1-yl)-6-methyl-4-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

295310

 
r & d systems
295310
10mg Each for $214.12

295350

 
r & d systems
295350
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 285.39
Quantity 10mg
Chemical Name or Material CyPPA
CAS 73029-73-9
Assay Percent Range >99%
Molecular Formula C16H23N5
Synonym N-Cyclohexyl-N-[2-(3, 5-dimethyl-pyrazol-1-yl)-6-methyl-4-pyrimidinamine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C