missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CYP4B1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00001580B01P
Description
Sequence: MVPSFLSLSFSSLGLWASGLILVLGFLKLIHLLLRRQTLAKAMDKFPGPPTHWLFGHALEIQETGSLDKVVSWAHQFPYAHPLWFGQFIGFLNIYEPDYAKAVYSRGDPKAPDVYDFFLQWIGRGLLVLEGPKWLQHRKLLTPGFHYDVLKPYVAVFTESTRIMLDKWEEKAREGKSFDIFCDVGHMALNTLMKCTFGRGDTGLGHSRDSSYYLAVSDLTLLMQQRLVSFQYHNDFIYWLTPHGRRFLRACQVAHDHTDQVIRERKAALQDEKVRKKIQNRRHLDFLDILLGARDEDDIKLSDADLRAEVDTFMFEGHDTTTSGISWFLYCMALYPEHQHRCREEVREILGDQDFFQWDDLGKMTYLTMCIKESFRLYPPVPQVYRQLSKPVTFVDGRSLPAGSLISMHIYALHRNSAVWPDPEVFDSLRFSTENASKRHPFAFMPFSAGPRNCIGQQFAMSEMKVVTAMCLLRFEFSLDPSRLPIKMPQLVLRSKNGFHLHLKPLGPGSGKSpecifications
CYP4B1 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC017758.1 | |
CYP4B1 | |
CYP4B1 (AAH17758.1, 1 a.a. ∼ 512 a.a) full-length human protein. | |
RUO | |
1580 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
cytochrome P450, family 4, subfamily B, polypeptide 1 | |
CYPIVB1/P-450HP | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |