Please login to your online account to display your discounted pricing

CYM 9484, Tocris™

Potent NPY Y2 receptor antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material CYM 9484
Assay Percent Range >98%
Molecular Formula C27H31N3O3S2
Formula Weight 509.63
Synonym N-[4-[(Dimethylamino)sulfonyl]phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinecarbothioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

460650

 
r & d systems
RDS460650
50mg Each for $976.15

460610

 
r & d systems
RDS460610
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 509.63
Quantity 50mg
Chemical Name or Material CYM 9484
Synonym N-[4-[(Dimethylamino)sulfonyl]phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinecarbothioamide
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
CAS 1383478-94-1
Molecular Formula C27H31N3O3S2
Safety and Handling

Store at Room Temperature