Please login to your online account to display your discounted pricing

CYM 50358 hydrochloride, Tocris™

Potent and selective S1P4 antagonist

$266.69 - $1,121.62

Chemical Identifiers

Chemical Name or Material CYM 50358 hydrochloride
Assay Percent Range >98%
Molecular Formula C20H18Cl2N2O2·HCl
Formula Weight 425.74
Synonym N-[(4-(Aminomethyl)-2, 6-dimethylphenyl]-5-(2, 5-dichlorophenyl)-2-furancarboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467910

 
r & d systems
RDS467910
10mg Each for $266.69

467950

 
r & d systems
RDS467950
50mg Each for $1,121.62
Description & Specifications

Specifications

Formula Weight 425.74
Quantity 10mg
Chemical Name or Material CYM 50358 hydrochloride
Molecular Formula C20H18Cl2N2O2·HCl
Synonym N-[(4-(Aminomethyl)-2, 6-dimethylphenyl]-5-(2, 5-dichlorophenyl)-2-furancarboxamide hydrochloride
Assay Percent Range >98%
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C