Please login to your online account to display your discounted pricing

CYM 50308, Tocris™

Potent and selective S1P4 agonist

$272.44 - $1,145.83

Chemical Identifiers

Chemical Name or Material CYM 50308
Assay Percent Range >98%
Molecular Formula C20H21F2N3O2S
Formula Weight 405.46
Synonym (2Z,5Z)-5-[[1-(2,4-Difluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]methylene]-2-[(2-methoxyethyl)imino]-3-methyl-4-thiazolidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467850

 
r & d systems
RDS467850
50mg Each for $1,145.83

467810

 
r & d systems
RDS467810
10mg Each for $272.44
Description & Specifications

Specifications

Formula Weight 405.46
Quantity 50mg
Assay Percent Range >98%
Synonym (2Z,5Z)-5-[[1-(2,4-Difluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]methylene]-2-[(2-methoxyethyl)imino]-3-methyl-4-thiazolidinone
Chemical Name or Material CYM 50308
Molecular Formula C20H21F2N3O2S
CAS 1345858-76-5
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C