Please login to your online account to display your discounted pricing

CYM 50260, Tocris™

Potent and selective S1P4 agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material CYM 50260
Assay Percent Range >98%
Molecular Formula C14H11Cl3FNO2
Formula Weight 350.6
Synonym 2-Chloro-3-[2-(2, 4-dichlorophenoxy)ethoxy]-6-(fluoromethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467710

 
r & d systems
RDS467710
10mg Each for $205.70

467750

 
r & d systems
RDS467750
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 350.6
Quantity 10mg
Assay Percent Range >98%
CAS 1355026-60-6
Synonym 2-Chloro-3-[2-(2, 4-dichlorophenoxy)ethoxy]-6-(fluoromethyl)pyridine
Recommended Storage Store at -20°C
Chemical Name or Material CYM 50260
Molecular Formula C14H11Cl3FNO2
Safety and Handling

Store at -20°C