Please login to your online account to display your discounted pricing

Cyclothiazide, Tocris

AMPA selective desensitization inhibitor

$100.81 - $427.46

Chemical Identifiers

Chemical Name or Material Cyclothiazide
Assay Percent Range >98%
Molecular Formula C14H16ClN3O4S2
Formula Weight 389.87
Synonym 6-Chloro-3,4-dihydro-3-(5-norbornen-2-yl)-2H-1,2,4-benzothiazidiazine-7-sulfonamide-1,1-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

071350

 
r & d systems
RDS071350
50mg Each for $427.46

071310

 
r & d systems
RDS071310
10mg Each for $100.81
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 389.87
Chemical Name or Material Cyclothiazide
Recommended Storage Store at Room Temperature
Synonym 6-Chloro-3,4-dihydro-3-(5-norbornen-2-yl)-2H-1,2,4-benzothiazidiazine-7-sulfonamide-1,1-dioxide
CAS 2259-96-3
Assay Percent Range >98%
Molecular Formula C14H16ClN3O4S2
Safety and Handling

Store at Room Temperature