missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopropylcarboxamidine hydrochloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2018514

 View more versions of this product

Catalog No. AAB2018514



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopropylcarboxamidine hydrochloride
122°C to 123°C
MFCD00053010
4507255
cyclopropanecarboximidamide hydrochloride, cyclopropane-1-carboximidamide hydrochloride, cyclopropanecarboxamidine hydrochloride, cyclopropylcarbamidine hydrochloride, cyclopropanecarboxamidine hcl, cyclopropyl carbamidine hydrochloride, clcyopropylcarbamidine hydrochloride, cyclopropylcarboxamidine hydrochloride, cyclopropyl carbamidine hcl, cyclopropanecarboximidamide, monohydrochloride
[H+].[Cl-].NC(=N)C1CC1
120.58
120.59
57297-29-7
C4H9ClN2
25 g
Hygroscopic
JRYOZJIRAVZGMV-UHFFFAOYSA-N
cyclopropanecarboximidamide;hydrochloride
2781916
97%

Chemical Identifiers

57297-29-7
120.58
JRYOZJIRAVZGMV-UHFFFAOYSA-N
2781916
C4H9ClN2
MFCD00053010
cyclopropanecarboximidamide hydrochloride, cyclopropane-1-carboximidamide hydrochloride, cyclopropanecarboxamidine hydrochloride, cyclopropylcarbamidine hydrochloride, cyclopropanecarboxamidine hcl, cyclopropyl carbamidine hydrochloride, clcyopropylcarbamidine hydrochloride, cyclopropylcarboxamidine hydrochloride, cyclopropyl carbamidine hcl, cyclopropanecarboximidamide, monohydrochloride
[H+].[Cl-].NC(=N)C1CC1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only