missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopropylamine, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC263685000

 View more versions of this product

Catalog No. AC263685000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclopropylamine
0.8240g/mL
49.0°C to 50.0°C
97.5% min. (GC)
C3H5NH2
12,3
1.4190 to 1.4220
500mL
HTJDQJBWANPRPF-UHFFFAOYSA-N
C1CC1N
57.09
CHEBI:34660
98%
765-30-0
-50.0°C
−25°C
C3H7N
MFCD00001301
Glass bottle
8pr8xth1x1, aminocyclopropane, cyclopropanamin, cycloproplyamine, cyclopropyamine, cyclopropyl amine, cyclopropylamin, cyclopropylamine, cylopropylamine, unii-8pr8xth1x1
Solubility in water: soluble. Other solubilities: soluble in ethanol
0.824
cyclopropanamine
69828
57.09

Chemical Identifiers

765-30-0
57.09
HTJDQJBWANPRPF-UHFFFAOYSA-N
69828
cyclopropanamine
C3H7N
MFCD00001301
8pr8xth1x1, aminocyclopropane, cyclopropanamin, cycloproplyamine, cyclopropyamine, cyclopropyl amine, cyclopropylamin, cyclopropylamine, cylopropylamine, unii-8pr8xth1x1
CHEBI:34660
C1CC1N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-142-2

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only