missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopropylacetylene, 97%

Manufacturer:  Alfa Aesar™ H5344022

 View more versions of this product

Catalog No. AAH5344022



Description

Description

Applications
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

Cyclopropylacetylene
−17°C (−1°F)
51°C
6746-94-7
MFCD02181090
cyclopropane, ethynyl
NPTDXPDGUHAFKC-UHFFFAOYSA-N
ethynylcyclopropane
138823
97%
0.781
1.429
100g
C5H6
UN3295
Slightly soluble in water.
C#CC1CC1
66.103
66.1

Chemical Identifiers

6746-94-7
66.103
NPTDXPDGUHAFKC-UHFFFAOYSA-N
138823
C#CC1CC1
C5H6
MFCD02181090
cyclopropane, ethynyl
ethynylcyclopropane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H318-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Causes skin irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P310-P332+P313-P363-P370+P378q-P501c

H225-H315-H318

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents