missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 443780010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclopropylacetic acid | |
1.1400g/mL | |
93°C | |
Glass bottle | |
1 g | |
cyclopropylacetic acid, cyclopropaneacetic acid, cyclopropyl acetic acid, cyclopropyl-acetic acid, carboxymethyl cyclopropane, 2-cyclopropyl-acetic acid, 2-cyclopropylethanoic acid, acetic cyclopropyl acid, pubchem19703, 2-cyclopropyl acetic acid | |
C1CC1CC(=O)O | |
100.12 | |
100.12 |
5239-82-7 | |
60.0°C to 61.0°C (29.0 mmHg) | |
97% | |
C5H8O2 | |
1.14 | |
KVVDRQDTODKIJD-UHFFFAOYSA-N | |
2-cyclopropylacetic acid | |
138440 | |
97% |
Chemical Identifiers
5239-82-7 | |
100.12 | |
cyclopropylacetic acid, cyclopropaneacetic acid, cyclopropyl acetic acid, cyclopropyl-acetic acid, carboxymethyl cyclopropane, 2-cyclopropyl-acetic acid, 2-cyclopropylethanoic acid, acetic cyclopropyl acid, pubchem19703, 2-cyclopropyl acetic acid | |
2-cyclopropylacetic acid |
C5H8O2 | |
KVVDRQDTODKIJD-UHFFFAOYSA-N | |
138440 | |
C1CC1CC(=O)O |
Safety and Handling
GHS Signal Word: Danger
RUO – Research Use Only