Please login to your online account to display your discounted pricing

Cyclopropyl(trimethylsilyl)acetylene, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C8H14Si
Linear Formula C3H5CCSi(CH3)3
Formula Weight 138.28
MDL Number MFCD00671355
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387520050

  SDS
thermo fisher scientific
AC387520050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC387520010

  SDS
thermo fisher scientific
AC387520010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.0%
Physical Form Liquid
Formula Weight 138.28
Quantity 5g
Color Clear colorless to yellow
Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Molecular Formula C8H14Si
Linear Formula C3H5CCSi(CH3)3
MDL Number MFCD00671355
CAS 81166-84-9
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97+%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.