Learn More
Cyclopropyl(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5348714
Description
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.
Solubility
Sparingly Soluble (0.069 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
83°C (80mmHg) | |
81166-84-9 | |
C8H14Si | |
UN1993 | |
cyclopropyl trimethylsilyl acetylene, cyclopropylethynyl trimethylsilane, 2-cyclopropylethynyl trimethylsilane, cyclopropylethynyl trimethyl silane, silane, cyclopropylethynyl trimethyl, cyclopropane, 2-trimethylsilyl ethynyl, silane, cyclopropylethynyl trimethyl-9ci, timtec-bb sbb009013, trimethylsilylethynylcyclopropane, cyclopropylethynyl-trimethylsilane | |
IYQSVLJDYLPMCX-UHFFFAOYSA-N | |
(2-cyclopropylethynyl)trimethylsilane | |
2758018 | |
97% |
Cyclopropyl(trimethylsilyl)acetylene | |
25 g | |
MFCD00671355 | |
25°C (77°F) | |
Sparingly Soluble (0.069g/L) (25°C). | |
C[Si](C)(C)C#CC1CC1 | |
138.29 | |
138.28 | |
1.454 |
Chemical Identifiers
81166-84-9 | |
138.29 | |
IYQSVLJDYLPMCX-UHFFFAOYSA-N | |
2758018 |
C8H14Si | |
MFCD00671355 | |
cyclopropyl trimethylsilyl acetylene, cyclopropylethynyl trimethylsilane, 2-cyclopropylethynyl trimethylsilane, cyclopropylethynyl trimethyl silane, silane, cyclopropylethynyl trimethyl, cyclopropane, 2-trimethylsilyl ethynyl, silane, cyclopropylethynyl trimethyl-9ci, timtec-bb sbb009013, trimethylsilylethynylcyclopropane, cyclopropylethynyl-trimethylsilane | |
C[Si](C)(C)C#CC1CC1 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only