missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopropyl(trimethylsilyl)acetylene, 97%

Manufacturer:  Alfa Aesar™ H5348706

 View more versions of this product

Catalog No. AAH5348706



Description

Description

Applications
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

Solubility
Sparingly Soluble (0.069 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Cyclopropyl(trimethylsilyl)acetylene
C8H14Si
UN1993
Sparingly Soluble (0.069g/L) (25°C).
C[Si](C)(C)C#CC1CC1
138.285
138.28
1.454
5g
81166-84-9
MFCD00671355
2-cyclopropylethynyl trimethylsilane
IYQSVLJDYLPMCX-UHFFFAOYSA-N
2-cyclopropylethynyl(trimethyl)silane
2758018
97%
83°C (80mmHg)
25°C (77°F)

Chemical Identifiers

81166-84-9
138.285
IYQSVLJDYLPMCX-UHFFFAOYSA-N
2758018
C[Si](C)(C)C#CC1CC1
C8H14Si
MFCD00671355
2-cyclopropylethynyl trimethylsilane
2-cyclopropylethynyl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents