Learn More
Cyclopropyl phenyl sulfide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1232103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclopropyl phenyl sulfide | |
1.045 | |
91°C (195°F) | |
C9H10S | |
MFCD00009684 | |
UN3334 | |
cyclopropyl phenyl sulfide, cyclopropyl phenyl sulfane, cyclopropylsulfanyl benzene, benzene, cyclopropylthio, cyclopropylphenylsulfide, cyclopropyl phenyl sulphide, unii-72c99lp48p, cyclopropylsulfanyl-benzene, pubchem10736, cyclopropylphenyl sulfide | |
C1CC1SC1=CC=CC=C1 | |
150.24 | |
150.24 |
14633-54-6 | |
61°C to 62°C (1 mmHg) | |
Stench | |
1.581 | |
1 g | |
2075701 | |
YIBKCPJOFAUAKY-UHFFFAOYSA-N | |
cyclopropylsulfanylbenzene | |
123348 | |
≥98% |
Chemical Identifiers
14633-54-6 | |
150.24 | |
YIBKCPJOFAUAKY-UHFFFAOYSA-N | |
123348 | |
C1CC1SC1=CC=CC=C1 |
C9H10S | |
MFCD00009684 | |
cyclopropyl phenyl sulfide, cyclopropyl phenyl sulfane, cyclopropylsulfanyl benzene, benzene, cyclopropylthio, cyclopropylphenylsulfide, cyclopropyl phenyl sulphide, unii-72c99lp48p, cyclopropylsulfanyl-benzene, pubchem10736, cyclopropylphenyl sulfide | |
cyclopropylsulfanylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only