missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopropyl methyl ketone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC111711000
Specifications
Cyclopropyl methyl ketone | |
21°C | |
Glass bottle | |
0.898 | |
0.2% max. | |
Colorless to Yellow | |
100mL | |
97.5% min. (GC) | |
C3H5COCH3 | |
07,7 | |
1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, acetylcyclopropane, cyclopropyl methyl ketone, cyclopropylethanone, cyclopropylmethylketone, ethanone, 1-cyclopropyl, ketone, cyclopropyl methyl, methyl cyclopropyl ketone, unii-n27yy1xcfh | |
HVCFCNAITDHQFX-UHFFFAOYSA-N | |
1-cyclopropylethanone | |
13004 | |
Liquid |
0.8980g/mL | |
Authentic | |
1.4220 to 1.4260 | |
0.65 mPa.s (20°C) | |
111.0°C | |
-68.0°C | |
765-43-5 | |
C5H8O | |
MFCD00001297 | |
01,676 | |
Solubility in water: 185g/L (20°C). Other solubilities: soluble in alcohols,chloride solvents | |
CC(=O)C1CC1 | |
84.12 | |
84.12 | |
98% |
Chemical Identifiers
765-43-5 | |
84.12 | |
HVCFCNAITDHQFX-UHFFFAOYSA-N | |
13004 | |
CC(=O)C1CC1 |
C5H8O | |
MFCD00001297 | |
1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, acetylcyclopropane, cyclopropyl methyl ketone, cyclopropylethanone, cyclopropylmethylketone, ethanone, 1-cyclopropyl, ketone, cyclopropyl methyl, methyl cyclopropyl ketone, unii-n27yy1xcfh | |
1-cyclopropylethanone |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
EINECSNumber : 212-146-4