Please login to your online account to display your discounted pricing

Cyclopropyl Methyl Carbinol 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  111700250

 View more versions of this product

Catalog No. AC111700250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 86.13
Assay ≥95%
Physical Form Liquid
Quantity 25g
Color Clear colorless
TSCA TSCA
EINECS Number 212-145-9
Beilstein 06, I, 4
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Refractive Index 1.4300 to 1.4320 (20°C, 589nm)
MDL Number MFCD00001300
Name Note 96%
Chemical Name or Material Cyclopropyl methyl carbinol
Assay Percent Range 96%
Molecular Formula C5H10O
Linear Formula C3H5CH(CH3)OH
CAS 765-42-4
Infrared Spectrum Authentic
Water 1% max.
Packaging Glass bottle
Synonym α-Methylcyclopropanemethanol
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclopropyl methyl carbinol
Name Note 96%
CAS 765-42-4
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C5H10O
Linear Formula C3H5CH(CH3)OH
Formula Weight 86.13
MDL Number MFCD00001300
Synonym α-Methylcyclopropanemethanol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents