Please login to your online account to display your discounted pricing

Cyclopropyl Isothiocyanate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375490050

 View more versions of this product

Catalog No. AC375490050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 99.16
Quantity 5g
Color Clear colorless to slightly yellow
Density 1.0700g/mL
Chemical Name or Material Cyclopropyl isothiocyanate
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C4H5NS
Linear Formula C3H5NCS
MDL Number MFCD00040874
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 56601-42-4

Specifications

Chemical Name or Material Cyclopropyl isothiocyanate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H5NS
Linear Formula C3H5NCS
Formula Weight 99.16
MDL Number MFCD00040874
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.

SDS & Safety Documents