Please login to your online account to display your discounted pricing

Cyclopropyl Isothiocyanate, 97%, ACROS Organics™

$57.69 - $57.69

Chemical Identifiers

Chemical Name or Material Cyclopropyl isothiocyanate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H5NS
Linear Formula C3H5NCS
Formula Weight 99.16
MDL Number MFCD00040874
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375490010

  SDS
thermo fisher scientific
AC375490010
Glass bottle 1g Each for $57.69

AC375490050

  SDS
thermo fisher scientific
AC375490050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 99.16
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.0700g/mL
Linear Formula C3H5NCS
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Molecular Formula C4H5NS
Assay Percent Range 97%
Packaging Glass bottle
CAS 56601-42-4
Infrared Spectrum Authentic
MDL Number MFCD00040874
Chemical Name or Material Cyclopropyl isothiocyanate
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.