missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopropyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1198506

 View more versions of this product

Catalog No. AAL1198506



Description

Description

Applications
Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications

Specifications

Cyclopropyl isothiocyanate
87°C to 88°C (35mmHg)
56601-42-4
MFCD00040874
3600974
JGFBQFKZKSSODQ-UHFFFAOYSA-N
isothiocyanatocyclopropane
92463
97%
57°C (134°F)
5g
C4H5NS
UN2929
cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato, cyclopropyl isothiocyanate
C1CC1N=C=S
99.151
99.16

Chemical Identifiers

56601-42-4
99.151
JGFBQFKZKSSODQ-UHFFFAOYSA-N
92463
C1CC1N=C=S
C4H5NS
MFCD00040874
cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato, cyclopropyl isothiocyanate
isothiocyanatocyclopropane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318-H226
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents