Learn More
Cyclopropyl cyanide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 111661000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopropyl cyanide | |
0.9100g/mL | |
32°C | |
97.5% min. (GC) | |
C4H5N | |
C3H5CN | |
09,4 | |
cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril | |
C1CC1C#N | |
67.09 | |
67.09 |
5500-21-0 | |
133.0°C to 135.0°C | |
Authentic | |
Glass bottle | |
1.4200 to 1.4220 | |
100g | |
0.91 | |
AUQDITHEDVOTCU-UHFFFAOYSA-N | |
cyclopropanecarbonitrile | |
79637 | |
98% |
Chemical Identifiers
5500-21-0 | |
67.09 | |
cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril | |
cyclopropanecarbonitrile |
C4H5N | |
AUQDITHEDVOTCU-UHFFFAOYSA-N | |
79637 | |
C1CC1C#N |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathin
GHS Signal Word: Danger
EINECSNumber : 226-836-8
RUO â Research Use Only