missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopropyl cyanide, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC111665000

 View more versions of this product

Catalog No. AC111665000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclopropyl cyanide
32°C
Colorless to Yellow
5500-21-0
Glass bottle
98%
1.4200 to 1.4220
0.91
AUQDITHEDVOTCU-UHFFFAOYSA-N
cyclopropanecarbonitrile
79637
Liquid
0.9100g/mL
133.0°C to 135.0°C
500g
Authentic
98%
C4H5N
C3H5CN
Cyclopropanecarbonitrile
C1CC1C#N
67.091
67.09

Chemical Identifiers

5500-21-0
67.091
Cyclopropanecarbonitrile
cyclopropanecarbonitrile
C4H5N
AUQDITHEDVOTCU-UHFFFAOYSA-N
79637
C1CC1C#N
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wash face, hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Danger

SDS Documents