Learn More
Cyclopropyl Carbinol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 296821000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopropyl carbinol | |
0.8900g/mL | |
35°C | |
98.5% min. (GC) | |
C4H8O | |
C3H5CH2OH | |
100mL | |
0.89 | |
Solubility in water: miscible. Other solubilities: miscible with ethanol,slightly soluble in carbon tetrachloride | |
OCC1CC1 | |
72.11 | |
4.04 mPa.s (20°C) | |
99% |
2516-33-8 | |
123°C to 124°C (738.0mmHg) | |
Authentic | |
Glass bottle | |
1.4310 to 1.433 | |
MFCD00001309 | |
06,4 | |
cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol | |
GUDMZGLFZNLYEY-UHFFFAOYSA-N | |
cyclopropylmethanol | |
75644 | |
72.11 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 219-735-5
RTECSNumber : GZ2212250
TSCA : TSCA
RUO â Research Use Only