missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopropyl Bromide 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 111640250

 View more versions of this product

Catalog No. AC111640250


 

Specifications Description & Specifications

Specifications

Cyclopropyl bromide
25mL
4333-56-6
Glass bottle
≥98.5%
1.4590 to 1.4610 (20°C, 589nm)
C3H5Br
MFCD00001271
Clear colorless
99%
Authentic
Liquid
99%
C3H5Br
120.98
Bromocyclopropane

Chemical Identifiers

4333-56-6
C3H5Br
120.98
Bromocyclopropane
≥98.5%
99%
C3H5Br
MFCD00001271
Cyclopropyl bromide, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 224-375-7

TSCA : TSCA