missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopropyl 4-fluorophenyl ketone, 98%

$28.12 - $117.88

Chemical Identifiers

CAS 772-31-6
Molecular Formula C10H9FO
Formula Weight 164.18
MDL Number MFCD00013728
Beilstein 2044844
Synonym (4-Fluorobenzoyl)cyclopropane
Chemical Name or Material Cyclopropyl 4-fluorophenyl ketone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1585206 SDS Alfa Aesar™
A1585206
5g Each for $28.12
AAA1585214 SDS Alfa Aesar™
A1585214
25g Each for $117.88
Specifications

Description

Applications
It is used in medicine. Indoles undergo smooth alkylation with cyclopropyl ketones in the presence of the CeCl3·7H2O/LiI reagent system in refluxing acetonitrile under neutral conditions to produce the corresponding C-3 substituted indole derivatives in good to high yields and with high selectivity.

Solubility
Not miscible with water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

772-31-6
164.18
2044844
Cyclopropyl 4-fluorophenyl ketone
C10H9FO
MFCD00013728
(4-Fluorobenzoyl)cyclopropane

Specifications

100°C (212°F)
5g
C10H9FO
MFCD00013728
(4-Fluorobenzoyl)cyclopropane
98%
119° to 120°C (13mmHg)
772-31-6
164.18
2044844
Cyclopropyl 4-fluorophenyl ketone