Please login to your online account to display your discounted pricing

Cyclopropyl(2-thienyl)methanone, 97%

$285.97 - $285.97

Chemical Identifiers

Chemical Name or Material Cyclopropyl 2-thienyl ketone
CAS 6193-47-1
CAS Min % 96.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8OS
Formula Weight 152.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC111740250

  SDS
thermo fisher scientific
111740250
25g Each for $285.97
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 152.22
Quantity 25g
Color Clear yellow
CAS Max % 100.0
Refractive Index 1.5835 to 1.5855 (20°C, 589nm)
CAS 6193-47-1
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Cyclopropyl 2-thienyl ketone
Molecular Formula C8H8OS
CAS Min % 96.5
Infrared Spectrum Authentic
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.