Learn More
Thermo Scientific Chemicals Cyclopropanesulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 439020010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopropanesulfonyl chloride | |
1.3800g/mL | |
60.0°C (2.0 Torr) | |
96% min. (GC) | |
C3H5ClO2S | |
Glass bottle | |
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride | |
PFWWSGFPICCWGU-UHFFFAOYSA-N | |
cyclopropanesulfonyl chloride | |
2760919 | |
Liquid |
139631-62-2 | |
Colorless to Yellow | |
96°C | |
Authentic | |
MFCD01631933 | |
1g | |
1.38 | |
ClS(=O)(=O)C1CC1 | |
140.58 | |
140.59 | |
97% |
Chemical Identifiers
139631-62-2 | |
140.58 | |
PFWWSGFPICCWGU-UHFFFAOYSA-N | |
2760919 | |
ClS(=O)(=O)C1CC1 |
C3H5ClO2S | |
MFCD01631933 | |
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride | |
cyclopropanesulfonyl chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
RUO â Research Use Only