missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopropanesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5550703

 View more versions of this product

Catalog No. AAH5550703



Description

Description

Applications
Cyclopropanesulfonamide is used as fine chemicals to the pharmaceutical, agrochemical, aroma, polymer and electronic industries.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Cyclopropanesulfonamide
1g
C3H7NO2S
cyclopropane sulfonamide
WMSPXQIQBQAWLL-UHFFFAOYSA-N
cyclopropanesulfonamide
15765418
97%
101°C to 106°C
154350-28-4
MFCD08705286
Soluble in water.
C1CC1S(=O)(=O)N
121.154
121.15

Chemical Identifiers

154350-28-4
121.154
WMSPXQIQBQAWLL-UHFFFAOYSA-N
15765418
C1CC1S(=O)(=O)N
C3H7NO2S
MFCD08705286
cyclopropane sulfonamide
cyclopropanesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents