Learn More
Cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 111621000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclopropanecarboxylic acid | |
1759-53-1 | |
Colorless to Yellow | |
182.0°C to 184.0°C | |
Authentic | |
Glass bottle | |
1.4370 to 1.4390 | |
MFCD00001287 | |
09, 4 | |
carboxycyclopropane, cyclopropylcarboxylic acid, cyclopropionic acid, cyclopropane carboxylic acid, cyclopropanecarboxylicacid, cpc-acid, cyclopropyl carboxylic acid, trimethylenecarboxylic acid, cyclopropane carboylic acid, cyclopropane-carboxylic acid | |
YMGUBTXCNDTFJI-UHFFFAOYSA-N | |
cyclopropanecarboxylic acid | |
15655 | |
86.09 | |
Liquid |
98% | |
17.0°C to 19.0°C | |
1.0850g/mL | |
71°C | |
97.5% min. (GC) | |
C4H6O2 | |
C3H5CO2H | |
100 g | |
1.085 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether, soluble in most common organic solvents | |
C1CC1C(=O)O | |
86.09 | |
CHEBI:23500 | |
98% |
Chemical Identifiers
1759-53-1 | |
86.09 | |
YMGUBTXCNDTFJI-UHFFFAOYSA-N | |
15655 | |
cyclopropanecarboxylic acid |
C4H6O2 | |
MFCD00001287 | |
carboxycyclopropane, cyclopropylcarboxylic acid, cyclopropionic acid, cyclopropane carboxylic acid, cyclopropanecarboxylicacid, cpc-acid, cyclopropyl carboxylic acid, trimethylenecarboxylic acid, cyclopropane carboylic acid, cyclopropane-carboxylic acid | |
CHEBI:23500 | |
C1CC1C(=O)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 217-162-5
RTECSNumber : GZ1100000
TSCA : TSCA
RUO – Research Use Only