Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopropanecarboxamide, 98%

$24.72 - $934.19

Chemical Identifiers

Chemical Name or Material Cyclopropanecarboxamide
CAS 6228-73-5
Molecular Formula C4H7NO
Formula Weight 85.11
MDL Number MFCD00013729
Beilstein 1924346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2187104

  SDS
alfa aesar™
B2187104
2g Each for $24.72

AAB2187109

  SDS
alfa aesar™
B2187109
10g Each for $63.43

AAB2187118

  SDS
alfa aesar™
B2187118
50g Each for $238.07

AAB2187130

  SDS
alfa aesar™
B2187130
250g Each for $934.19
Description & Specifications

Specifications

Formula Weight 85.11
Molecular Formula C4H7NO
MDL Number MFCD00013729
CAS 6228-73-5
TSCA TSCA
Quantity 2g
Chemical Name or Material Cyclopropanecarboxamide
Assay 98%
Melting Point 122° to 126°C
Beilstein 1924346

Applications
Cyclopropanecarboxamide is used in the preparation of 2-(cyclopropanecarbonyl-amino)-but-2-enoic acid by reacting with 2-oxo-butyric acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases, strong reducing agents, acid chlorides and acid anhydrides.