missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopiazonic acid, 99+%

Inhibitor of the sarco/endoplasmic reticulum calcium pump, calcium-ATPase

Manufacturer:  Alfa Aesar™ J61594MF

 View more versions of this product

Catalog No. AAJ61594MF



Description

Description

Applications
The use of Cyclopiazonic acid on Langendorff-perfused murine hearts displayed anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis. It is also used as an inhibitor of endoplasmic reticulum Ca2+-ATPase.

Solubility
Soluble in DMSO (100 mM), ethanol (5 mM), DMF (~20 mg/ml), and 1:1 DMSO:PBS(pH 7.2) (0.5 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Keep in a dry place. Keep away from strong oxidizing agents.
Specifications

Specifications

Cyclopiazonic acid
50mg
C20H20N2O3
UN2811
SZINUGQCTHLQAZ-AFAVFJNCSA-N
336.391
336.39
≥99%
White
18172-33-3
MFCD00167445
cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
54711281
Powder

Chemical Identifiers

18172-33-3
336.391
SZINUGQCTHLQAZ-AFAVFJNCSA-N
54711281
C20H20N2O3
MFCD00167445
cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O
Safety and Handling

Safety and Handling

GHS H Statement
H300-H361
Fatal if swallowed.
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UY8587000

Recommended Storage : −30° to −10°C

Documents