Learn More
Cyclophosphamide monohydrate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 203960250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclophosphamide monohydrate | |
>110°C | |
96% min. (ELSD) (HPLC) | |
C7H17Cl2N2O3P | |
25g | |
cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated | |
PWOQRKCAHTVFLB-UHFFFAOYNA-N | |
N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate | |
22420 | |
279.09 |
6055-19-2 | |
Authentic | |
Glass bottle | |
MFCD00149395 | |
15, 2743 | |
Solubility in water: 40g/L. Other solubilities: slightly soluble in alcohol,benzene,ethylene,glycol,carbon tetrachloride and dioxane,sparingly soluble in ether and acetone | |
O.ClCCN(CCCl)P1(=O)NCCCO1 | |
279.10 | |
CHEBI:4026 | |
97% |
Safety and Handling
GHS H Statement
May cause genetic defects.
May cause cancer.
May damage the unborn child.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Wash face,hands and any exposed skin thoroughly after
GHS Signal Word: Danger
RUO â Research Use Only