Learn More
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203960050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclophosphamide monohydrate | |
49.0°C to 53.0°C | |
>110°C | |
96% min. (ELSD) (HPLC) | |
C7H17Cl2N2O3P | |
5 g | |
cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated | |
PWOQRKCAHTVFLB-UHFFFAOYNA-N | |
N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate | |
22420 | |
279.09 | |
Crystalline Powder or Crystals |
6055-19-2 | |
White | |
Authentic | |
Glass bottle | |
MFCD00149395 | |
15, 2743 | |
Solubility in water: 40g/L. Other solubilities: slightly soluble in alcohol,benzene,ethylene,glycol,carbon tetrachloride and dioxane,sparingly soluble in ether and acetone | |
O.ClCCN(CCCl)P1(=O)NCCCO1 | |
279.10 | |
CHEBI:4026 | |
97% |
Chemical Identifiers
6055-19-2 | |
279.10 | |
PWOQRKCAHTVFLB-UHFFFAOYNA-N | |
22420 | |
N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate |
C7H17Cl2N2O3P | |
MFCD00149395 | |
cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated | |
CHEBI:4026 | |
O.ClCCN(CCCl)P1(=O)NCCCO1 |
Safety and Handling
GHS H Statement
May cause genetic defects.
May cause cancer.
May damage the unborn child.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Wash face,hands and any exposed skin thoroughly after
GHS Signal Word: Danger
RUO – Research Use Only