Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclophosphamide monohydrate, 97+%

Chemical Identifiers

Chemical Name or Material Cyclophosphamide monohydrate
CAS 6055-19-2
Molecular Formula C7H15Cl2N2O2P·H2O
Formula Weight 279.10 (261.09 Anhydrous)
MDL Number MFCD00149395
UN Number UN3464
Beilstein 8167897
Merck Index 14,2747
Synonym Bis(2-chloroethyl)phosphoramide cyclic propanolamide ester; Cyclophosphane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1150803

  SDS
alfa aesar™
L1150803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1150806

  SDS
alfa aesar™
L1150806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1150814

  SDS
alfa aesar™
L1150814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 279.10 (261.09 Anhydrous)
Chemical Name or Material Cyclophosphamide monohydrate
MDL Number MFCD00149395
Assay ≥97%
Beilstein 8167897
Recommended Storage 2° to 8°C
Flash Point 113°C (235°F)
Molecular Formula C7H15Cl2N2O2P·H2O
Melting Point 51° to 53°C
Synonym Bis(2-chloroethyl)phosphoramide cyclic propanolamide ester; Cyclophosphane
Health Hazard 3 GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
RTECS Number RP6157750
Odor Odorless
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
Health Hazard 2 GHS H Statement
H301-H340-H350-H360
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Merck Index 14,2747
UN Number UN3464
CAS 6055-19-2

Applications
Cyclophosphamide monohydrate is used to treat cancers, autoimmune disorders and AL amyloidosis. It is a nitrogen mustard alkylating agent and reacts with deoxyribonucleic acid (DNA) to get its alkylated derivative. It is also involved in cancer research as a tool that induces apoptosis in tumor cells.

Solubility
Soluble in water. Slightly soluble ethanol, benzene, ethylene glycol, carbon tetrachloride, dioxane, ether, acetone, alcohol and chloroform.

Notes
Special handling precautions required. View MSDS prior to purchase, available online at www.alfa.comStore in cool place. Moisture and light senstive. Incompatible with strong oxidizing agents, strong acids and strong bases.

Safety and Handling

Danger

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H340-H350-H360
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.

2° to 8°C