missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentyltriphenylphosphonium bromide, 98%

Manufacturer:  Alfa Aesar™ A1464730

 View more versions of this product

Catalog No. AAA1464730



Specifications

Specifications

Cyclopentyltriphenylphosphonium bromide
250g
C23H24BrP
3580193
WZYWSVSFFTZZPE-UHFFFAOYSA-M
cyclopentyl(triphenyl)phosphanium;bromide
2735896
98%
262°C to 266°C
7333-52-0
MFCD00051880
cyclopentyltriphenylphosphonium bromide, cyclopentyl triphenyl phosphanium bromide, phosphonium, cyclopentyltriphenyl-, bromide, cyclopentyltriphenylphosphanium bromide, cyclopentyl triphenyl phosphonium bromide, cyclopentyltriphenylphosphoniumbromide, cyclopentyl triphenylphosphonium bromide, phosphonium,cyclopentyltriphenyl-,bromide 1:1, phosphonium,cyclopentyltriphenyl-, bromide 1:1
C1CCC(C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
411.323
411.33

Chemical Identifiers

7333-52-0
411.323
WZYWSVSFFTZZPE-UHFFFAOYSA-M
2735896
C1CCC(C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C23H24BrP
MFCD00051880
cyclopentyltriphenylphosphonium bromide, cyclopentyl triphenyl phosphanium bromide, phosphonium, cyclopentyltriphenyl-, bromide, cyclopentyltriphenylphosphanium bromide, cyclopentyl triphenyl phosphonium bromide, cyclopentyltriphenylphosphoniumbromide, cyclopentyl triphenylphosphonium bromide, phosphonium,cyclopentyltriphenyl-,bromide 1:1, phosphonium,cyclopentyltriphenyl-, bromide 1:1
cyclopentyl(triphenyl)phosphanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents