Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentyltrichlorosilane, 97%

Chemical Identifiers

Chemical Name or Material Cyclopentyltrichlorosilane
CAS 14579-03-4
Molecular Formula C5H9Cl3Si
Formula Weight 203.57
MDL Number MFCD00236508
UN Number UN2987
Beilstein 2935230
Synonym Trichlorocyclopentylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1644114

  SDS
alfa aesar™
L1644114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1644122

  SDS
alfa aesar™
L1644122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.57
Molecular Formula C5H9Cl3Si
CAS 14579-03-4
Chemical Name or Material Cyclopentyltrichlorosilane
Synonym Trichlorocyclopentylsilane
Health Hazard 1 Danger
TSCA TSCA
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
UN Number UN2987
MDL Number MFCD00236508
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Beilstein 2935230
Flash Point 68°C (154°F)
Boiling Point 178° to 179°C
Quantity 25g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
Odor Acrid
Assay 97%
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.