Learn More
Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427458000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopentylmagnesium bromide | |
33240-34-5,60-29-7 | |
35°C | |
AcroSeal™ Glass Bottle | |
MFCD00209569 | |
0.996 | |
JMPXNLAYOWCGCW-UHFFFAOYSA-M | |
magnesium;cyclopentane;bromide | |
118418 |
2.0M solution in diethyl ether | |
0.9960g/mL | |
-38°C | |
C5H9BrMg | |
800mL | |
cyclopentylmagnesium bromide, grignard reagent, magnesium, bromocyclopentyl, magnesium, bromocyclopentyl, bromo cyclopentyl magnesium, bromo cyclopentyl magnesium, cydopentylmagnesium bromide, cydopentylmagnesium bromide, pzojujcaldffhc-uhfffaoysa-m, cyclopentylmagnesium bromide solution, 2.0 m in diethyl ether, cyclopentylmagnesium bromide solution, 2.0 m in diethyl ether | |
C1CC[CH-]C1.[Mg+2].[Br-] | |
173.33 | |
173.33 |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Extremely flammable liquid and vapor. In contact with water releases flammable gas. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.
GHS Signal Word: Danger
EINECSNumber : 251-420-8
RUO â Research Use Only