Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentylboronic acid, 95%

$86.51 - $443.31

Chemical Identifiers

Chemical Name or Material Cyclopentylboronic acid
CAS 63076-51-7
Molecular Formula C5H11BO2
Formula Weight 113.95
MDL Number MFCD01074541
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2708103

 
alfa aesar™
H2708103
1g Each for $86.51

AAH2708109

 
alfa aesar™
H2708109
10g Each for $443.31
Description & Specifications

Specifications

Formula Weight 113.95
Chemical Name or Material Cyclopentylboronic acid
Assay 95%
Molecular Formula C5H11BO2
MDL Number MFCD01074541
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
CAS 63076-51-7
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.