Learn More
Cyclopentylamine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 111590250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclopentylamine | |
1003-03-8 | |
106°C to 108°C | |
Authentic | |
Glass bottle | |
1.4482 to 1.4502 | |
MFCD00001380 | |
12,4 | |
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine | |
NISGSNTVMOOSJQ-UHFFFAOYSA-N | |
cyclopentanamine | |
2906 | |
99+% |
99+% | |
0.8600g/mL | |
17°C | |
99% min. (GC) | |
C5H11N | |
C5H9NH2 | |
25 g | |
0.86 | |
Solubility in water: completely miscible with water | |
NC1CCCC1 | |
85.15 | |
85.15 | |
Liquid |
Chemical Identifiers
1003-03-8 | |
85.15 | |
NISGSNTVMOOSJQ-UHFFFAOYSA-N | |
2906 | |
NC1CCCC1 |
C5H11N | |
MFCD00001380 | |
cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine | |
cyclopentanamine |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 213-697-3
RTECSNumber : GY8452000
TSCA : TSCA
RUO – Research Use Only