Please login to your online account to display your discounted pricing

Cyclopentylacetylene, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381310010

 View more versions of this product

Catalog No. AC381310010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 94.16
Physical Form Liquid
Quantity 1g
Color Clear yellow
Chemical Name or Material Cyclopentylacetylene
Linear Formula CHCC5H9
CAS 930-51-8
Molecular Formula C7H10
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Infrared Spectrum Authentic
Refractive Index 1.447 to 1.449 (20°C, 589nm)
Packaging Glass bottle
Synonym Ethynylcyclopentane
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00013744
Assay Percent Range 95%

Specifications

Chemical Name or Material Cyclopentylacetylene
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H10
Linear Formula CHCC5H9
Formula Weight 94.16
MDL Number MFCD00013744
Synonym Ethynylcyclopentane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents